Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Antibody detects endogenous levels of total PTP4A3.
The protein encoded by this gene belongs to a small class of prenylated protein tyrosine phosphatases (PTPs). PTPs are cell signaling molecules that play regulatory roles in a variety of cellular processes. This class of PTPs contain a PTP domain and a characteristic C-terminal prenylation motif. Studies of this class of PTPs in mice demonstrated that they were prenylated proteins in vivo, which suggested their association with cell plasma membrane. Overexpression of this gene in mammalian cells was reported to inhibit angiotensin-II induced cell calcium mobilization and promote cell growth. Two alternatively spliced variants exist. PRL-3 expression was found to be up-regulated in human liver carcinoma compared with normal liver. Its expression in colorectal cancers may contribute to the establishment of liver metastasis. In a different study, PRL-3 is found to be expressed in tumor metastasis and vasculature, regardless of the tumor source.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HPRL-3; potentially prenylated protein tyrosine phosphatase; PRL-3; PRL-R; protein tyrosine phosphatase 4a3; Protein tyrosine phosphatase type IVA 3; Protein-tyrosine phosphatase 4a3; Protein-tyrosine phosphatase of regenerating liver 3
Gene Aliases: AV088979; pPtp4a3; PRL-3; PRL-R; PRL3; PTP4A3
UniProt ID: (Human) O75365, (Mouse) Q9D658
Entrez Gene ID: (Human) 11156, (Mouse) 19245, (Rat) 362930
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support