Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MEVSCGQAES SEKPNAEDMT SKDYYFDSYA HFGIHEEMLK DEVRTLTYRN SMFHNRHLFK DKVVLDVGSG TGILCMFAAK AGARKVIGIE CSSISDYAVK IVKANKLDHV VTIIKGKVEE VELPVEKVDI IISEWMGYCL FYESMLNTVL YARDKWLAPD GLIFPDRATL YVTAIEDRQY KDYKIHWWEN VYGFDMSCIK DVAIKEPLVD VVDPKQLVTN ACLIKEVDIY TVKVEDLTFT SPFCLQVKRN DYVHALVAYF NIEFTRCHKR TGFSTSPESP YTHWKQTVFY MEDYLTVKTG EEIFGTIGMR PNAKNNRDLD FTIDLDFKGQ LCELSCSTDY RMR
PRMT1 is a member of the protein arginine N-methyltransferase (PRMT) family. Arginine methylation is an irreversible post translational modification which has been implicated in RNA processing and trafficking, receptor-mediated signaling, and transcription. At least three types of PRMT enzymes have been identified in mammalian cells. These enzymes have been shown to have essential regulatory functions by methylation of key proteins in several fundamental areas. These proteins include nuclear proteins, IL enhancer binding factor, nuclear factors, cell cycle proteins, signal transduction proteins, apoptosis proteins, and viral proteins. The mammalian PRMT family currently consists of 7 members that share two large domains of homology. Increased expression of PRMT1 may play a role in various types of cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: arginine N-methyltransferase 1; heterogeneous nuclear ribonucleoprotein methyltransferase 1-like 2; heterogeneous nuclear ribonucleoproteins methyltransferase-like 2; Histone-arginine N-methyltransferase PRMT1; HMT1 (hnRNP methyltransferase, S. cerevisiae)-like 2; Interferon receptor 1-bound protein 4; Protein arginine N-methyltransferase 1
Gene Aliases: 6720434D09Rik; ANM1; AW214366; HCP1; HMT2; HRMT1L2; IR1B4; Mrmt1; PRMT1
UniProt ID: (Human) Q99873, (Mouse) Q9JIF0, (Rat) Q63009
Entrez Gene ID: (Human) 3276, (Mouse) 15469, (Rat) 60421
Molecular Function: protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support