Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ESLTDKVILD VGCGTGIISL FCAHYARPRA VYAVEASEMA QHTGQLVLQN GFADIITVYQ QKVEDVVLPE KVDVLVSEWM GTCLLFEFMI ESILYARDAW LKEDGVIWPT MAALHLVPCS ADKDYRSKVL FWDNAYEFNL SALKSLAVKE FFSKPKYNHI LKPEDCLSEP CTILQLDMRT VQISDLETLR GELRFDIRKA GTLHGFTAWF SVHFQSLQEG QPPQVLSTGP FHPTTHWKQT LFMMDDPVPV HTGDVVTGSV VLQRNPVWIR HMSVALSWAV TSRQDPTSQK VGEKVFPIWR
Arginine methyltransferase that methylates the guanidino nitrogens of arginyl residues in proteins such as STAT3, FBL, histone H4. Acts as a coactivator (with NCOA2) of the androgen receptor (AR)-mediated transactivation. Acts as a coactivator (with estrogen) of estrogen receptor (ER)-mediated transactivation. Enhances PGR, PPARG, RARA-mediated transactivation. May inhibit NF-kappa-B transcription and promote apoptosis. Represses E2F1 transcriptional activity (in a RB1-dependent manner). May be involved in growth regulation. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Histone-arginine N-methyltransferase PRMT2; HMT1 (hnRNP methyltransferase, S. cerevisiae)-like 1; HMT1 hnRNP methyltransferase-like 1; MGC111373; PRMT2 alpha; PRMT2 beta; PRMT2 gamma; Protein arginine N-methyltransferase 2
Gene Aliases: AI504737; HMT1; HRMT1L1; PRMT2
UniProt ID: (Human) P55345
Entrez Gene ID: (Human) 3275, (Mouse) 15468, (Rat) 499420
Molecular Function: protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support