Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Western blot analysis using this antibody detected a major band around 42 kDa in Hela cells.
Immunogen sequence: MIADRVRTD AYRLGILRNW AALRGKTVLD VGAGTGILSI FCAQAGARRV YAVEASAIWQ QAREVVRFNG LEDRVHVLPG PVETVELPEQ VDAIVSEWMG YGLLHESMLS SVLHARTKWL KEGGLLLPAS AELFIAPISD QMLEWRLGFW SQVKQHYGVD MSCLEGFATR CLMGHSEIVV QGLSGEDVLA RPQRFAQLEL SRAGLEQELE AGVGGRFRCS CYGSAPMHGF AIWFQVTFPG GESEKPLVLS TSPFHPATHW KQALLYLNEP VQVEQDTDVS GEITLLPSRD NPRRLRVLLR YKVGDQEEKT KDFAMED (1-316 aa encoded by BC002729)
Protein arginine N-methyltransferase 6 (PRMT6) PRMT6 demonstrates type I PRMT activity, capable of forming both omega-N monomethylarginine and asymmetric dimethylarginine derivatives on the recombinant glycine- and arginine-rich substrates. cts as a transcriptional coactivator of a number of steroid hormone receptors including ESR1, ESR2, PGR and NR3C1. Promotes fasting-induced transcriptional activation of the gluconeogenic program through methylation of the CRTC2 transcription coactivator. May play a role in innate immunity against HIV-1 in case of infection by methylating and impairing the function of various HIV-1 proteins such as Tat, Rev and Nucleocapsid protein p7.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ10559; FLJ51477; Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 6; Histone-arginine N-methyltransferase PRMT6; HMT1 hnRNP methyltransferase-like 6; PRMT-6; Protein arginine N-methyltransferase 6
Gene Aliases: AW124876; BB233495; HRMT1L6; PRMT6
UniProt ID: (Human) Q96LA8, (Mouse) Q6NZB1
Entrez Gene ID: (Human) 55170, (Mouse) 99890
Molecular Function:
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support