Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody recognizes all of isoforms of PRMT8.
Immunogen sequence: RTLTYRNSM YHNKHVFKDK VVLDVGSGTG ILSMFAAKAG AKKVFGIECS SISDYSQKII KANHLDNIIT IFKGKVEEVE LPVEKVDIII SEWMGYCLFY ESMLNTVIFA RDKWLKPGGL MFPDRAALYV VAIEDRQYKD FKIHWWENVY GFDMTCIRDV AMKEPLVDIV DPKQVVTNAC LIKEVDIYTV KTEELSFTSA FCLQIQRNDY VHALVTYFNI EFTKCHKKMG FSTAPDAPYT HWKQTVFYLE DYLTVRRGEE IYGTISMKPN AKNVRDLDFT VDLDFKGQLC ETSVSNDYKM R (95-394 aa encoded by BC022458)
Arginine methylation is a widespread posttranslational modification mediated by arginine methyltransferases, such as PRMT8. Arginine methylation is involved in a number of cellular processes, including DNA repair, RNA transcription, signal transduction, protein compartmentalization, and possibly protein translation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: arginine methyltransferase 8; heterogeneous nuclear ribonucleoprotein methyltransferase-like 4; Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 4; HMT1 hnRNP methyltransferase-like 3; protein arginine N-methyltransferase 4; Protein arginine N-methyltransferase 8
Gene Aliases: HRMT1L3; HRMT1L4; PRMT8
UniProt ID: (Human) Q9NR22, (Mouse) Q6PAK3
Entrez Gene ID: (Human) 56341, (Mouse) 381813, (Rat) 688502
Molecular Function: protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support