Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 6
Positive test controls include: 293T, Mouse brain, Rat brain. The target is usually found in the following locations: Cell membrane, Cytoplasmic side, Lipid-anchor.
Immunogen sequence: VIFARDKWLK PGGLMFPDRA ALYVVAIEDR QYKDFKIHWW ENVYGFDMTC IRDVAMKEPL VDIVDPKQVV TNACLIKEVD IYTVKTEELS FTSAFCLQIQ RNDYVHALVT YFNIEFTKCH KKMGFSTAPD APYTHWKQTV FYLEDYLTVR RGEEIYGTIS MKPNAKNVRD LDFTVDLDFK G
Arginine methylation is a widespread posttranslational modification mediated by arginine methyltransferases, such as PRMT8. Arginine methylation is involved in a number of cellular processes, including DNA repair, RNA transcription, signal transduction, protein compartmentalization, and possibly protein translation.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: arginine methyltransferase 8; heterogeneous nuclear ribonucleoprotein methyltransferase-like 4; Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 4; HMT1 hnRNP methyltransferase-like 3; protein arginine N-methyltransferase 4; Protein arginine N-methyltransferase 8
Gene Aliases: HRMT1L3; HRMT1L4; PRMT8
UniProt ID: (Human) Q9NR22, (Mouse) Q6PAK3
Entrez Gene ID: (Human) 56341, (Mouse) 381813, (Rat) 688502
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support