Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: LGIPLDGTVC FGQLLGMCDH VSLALGQAGY VVYKSIPYGS LEEVIPYLIR
Sequence homology: Cow: 100%; Dog: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%
PRODH2 is similar to proline dehydrogenase (oxidase) 1, a mitochondrial enzyme which catalyzes the first step in proline catabolism. The function of this protein has not been determined.The protein encoded by this gene is similar to proline dehydrogenase (oxidase) 1, a mitochondrial enzyme which catalyzes the first step in proline catabolism. The function of this protein has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HsPOX1; Hydroxyproline dehydrogenase; HYPDH; Kidney and liver proline oxidase 1; Probable proline dehydrogenase 2; Probable proline oxidase 2; proline dehydrogenase (oxidase) 2
Gene Aliases: HSPOX1; HYPDH; PRODH2
UniProt ID: (Human) Q9UF12
Entrez Gene ID: (Human) 58510
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support