Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Samples: Mouse brain
Immunogen sequence: GGGRSRFSPQ GGQRGRPHSR RRHRTTFSPV QLEQLESAFG RNQYPDIWAR ESLARDTGLS EARIQVWFQN RRAKQRKQER SLLQPLAHLS PAAFSSFLPE S
PROP1 encodes a paired-like homeodomain transcription factor in the developing pituitary gland. Expression occurs prior to and is required for expression of pou domain transcription factor 1, which is responsible for pituitary development and hormone expression. Mutations in this gene have been associated with combined pituitary hormone deficiency-2 as well as deficiencies in luteinizing hormone, follicle-stimulating hormone, growth hormone, prolactin, and thyroid-stimulating hormone.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ames dwarf; Homeobox protein prophet of Pit-1; pituitary specific homeodomain factor; Pituitary-specific homeodomain factor; PROP-1; prophet of Pit-1
Gene Aliases: df; Prop-1; Prop1
UniProt ID: (Mouse) P97458
Entrez Gene ID: (Mouse) 19127
Molecular Function:
homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support