Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRVLGGRCGA LLACLLLVLP VSEANFLSKQ QASQVLVRKR RANSLLEETK QGNLERECIE ELCNKEEARE VFENDPETDY FYPKYLVCLR SFQTGLFTAA RQSTNAYPDL RSCVNAIPDQ CSPLPCNEDG YMSCKDGKAS FTCTCKPGWQ GEKCEFDINE CKDPSNINGG CSQICDNTPG SYHCSCKNGF VMLSNKKDCK DVDECSLKPS ICGTAVCKNI PGDFECECPE GYRYNLKSKS CEDIDECSEN MCAQLCVNYP GGYTCYCDGK KGFKLAQDQK SCEVVSVCLP LNLDTKYELL YLAEQFAGVV LYLKFRLPEI SRFSAEFDFR TYDSEGVILY AESIDHSAWL LIALRGGKIE VQLKNEHTSK ITTGGDVINN GLWNMVSVEE LEHSISIKIA KEAVMDINKP GPLFKPENGL LETKVYFAGF PRKVESELIK PINPRLDGCI RSWNLMKQGA SGIKEIIQEK QNKHCLVTVE KGSYYPGSGI AQFHIDYNNV SSAEGWHVNV TLNIRPSTGT GVMLALVSGN NTVPFAVSLV DSTSEKSQDI LLSVENTVIY RIQALSLCSD QQSHLEFRVN RNNLELSTPL KIETISHEDL QRQLAVLDKA MKAKVATYLG GLPDVPFSAT PVNAFYNGCM EVNINGVQLD LDEAISKHND IRAHSCPSVW KKTKNS
Protein S is a vitamin K-dependent, potent neutral anticoagulant plasma glycoprotein. Protein S has a molecular weight of approximately 70 kDa and consists of a single polypeptide chain. The concentration of protein S in human plasma is 25 µg/mL, with a half-life of approximately two days. About 40% of Protein S in plasma is in a free form, whereas 60% is complexed with C4b-binding Protein (C4BP). Only the free Protein S functions as a cofactor to antigen presenting cells (APC) in the APC-dependent degradation of Factor Va and Factor VIIIa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Preproprotein S; PROS 1; Protein S alpha; protein Sa; proteinS; PS 21; PS 22; PS 23; PS 24; PS 25; PS 26; PS26; vitamin K-dependent plasma protein S; Vitamin K-dependent protein S
Gene Aliases: AW214361; PROS; PROS1; PS21; PS22; PS23; PS24; PS25; PSA; THPH5; THPH6
UniProt ID: (Human) P07225, (Mouse) Q08761
Entrez Gene ID: (Human) 5627, (Mouse) 19128
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support