Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDILKSEILR KRQLVEDRNL LVENKKYFKR SELAKKEEEA YFERCGYKIQ PKEEDQKPLT SSNPVLELEL AEEKLPMTLS RQEVIRRLRE RGEPIRLFGE TDYDAFQRLR KIEILTPEVN KGLRNDLKAA LDKIDQQYLN EIVGGQEPGE EDTQNDLKVH EENTTIEELE ALGESLGKGD DHKDMDIITK FLKFLLGVWA KELNAREDYV KRSVQGKLNS ATQKQTESYL RPLFRKLRKR NLPADIKESI TDIIKFMLQR EYVKANDAYL QMAIGNAPWP IGVTMVGIHA RTGREKIFSK HVAHVLNDET QRKYIQGLKR LMTICQKHFP TDPSKCVEYN AL
PRPF18 (pre-mRNA-splicing factor 18), also known as HPRP18, is a 342 amino acid protein that localizes to nuclear speckles and plays a role in the second step of pre-mRNA splicing. A member of the PRP18 family, PRPF18 contains seven WD repeats and exists as two alternatively spliced isoforms which are encoded by a gene located on human chromosome 10p13. Chromosome 10 contains over 800 genes and 135 million nucleotides. PTEN is an important tumor suppressor gene located on chromosome 10 and, when defective, causes a genetic predisposition to cancer development known as Cowden syndrome. Other chromosome 10 associated disorders include Cockayne syndrome, tetrahydrobiopterin deficiency and trisomy 10.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPRP18; Pre-mRNA-splicing factor 18; PRP18 homolog; PRP18 pre-mRNA processing factor 18 homolog; PRP18 pre-mRNA processing factor 18 homolog(PRPF18)
Gene Aliases: HPRP18; PRP18; PRPF18
UniProt ID: (Human) Q99633
Entrez Gene ID: (Human) 8559
Molecular Function: RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support