Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ELHKSLRSL NNFCSQIGDD RPISYCHFSP NSKMLATACW SGLCKLWSVP DCNLLHTLRG HNTNVGAIVF HPKSTVSLDP KDVNLASCAA DGSVKLWSLD SDEPVADIEG HTVRVARVMW HPSGRFLGTT CYDRSWRLWD LEAQEEILHQ EGHSMGVYDI AFHQDGSLAG TGGLDAFGRV WDLRTGRCIM FLEGHLKEIY GINFSPNGYH IATGSGDNTC KVWDLRQRRC VYTIPAHQNL VTGVKFEPIH GNFLLTGAYD NTAKIWTHPG WSPLKTLAGH EGKVMGLDIS SDGQLIATCS YDRTFKLWMA E (213-522 aa encoded by BC001588)
The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. PRPF4 is 1 of several proteins that associate with U4 and U6 snRNPs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPrp4; PRP4 homolog; PRP4 pre-mRNA processing factor 4 homolog; PRP4/STK/WD splicing factor; U4/U6 small nuclear ribonucleoprotein Prp4; U4/U6 snRNP 60 kDa protein; WD splicing factor Prp4
Gene Aliases: 1600015H11Rik; AI874830; AW047464; bN189G18.1; HPRP4; HPRP4P; PRP4; Prp4p; PRPF4; RP70; SNRNP60
UniProt ID: (Human) O43172, (Mouse) Q9DAW6
Entrez Gene ID: (Human) 9128, (Mouse) 70052, (Rat) 298095
Molecular Function: RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support