Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Catalyzes the synthesis of phosphoribosylpyrophosphate (PRPP) that is essential for nucleotide synthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hCG_1643027; Phosphoribosyl pyrophosphate synthase 1-like 1; Phosphoribosyl pyrophosphate synthase III; phosphoribosylpyrophosphate synthetase subunit III; PRPS1-like 1; PRS-III; ribose-phosphate diphosphokinase catalytic chain III; Ribose-phosphate pyrophosphokinase 3; ribose-phosphate pyrophosphokinase III
Gene Aliases: PRPS1; PRPS1L1; PRPS3; PRPSL; PRS-III
UniProt ID: (Human) P21108
Entrez Gene ID: (Human) 221823
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support