Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • PRPS2 Antibodies

            Invitrogen

            PRPS2 Polyclonal Antibody

            3 Published Figures
            4 References
            View all (7) PRPS2 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite PRPS2 Polyclonal Antibody

            • Antibody Testing Data (2)
            • Published Figures (3)
            Application
            PRPS2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 5

            PRPS2 Antibody (PA5-42007) in IHC (P)

            Immunohistochemistry (paraffin-embedded) analysis of human kidney tissue using an anti-PRPS2 polyclonal antibody (Product # PA5-42007).
            View Product

            PRPS2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            PRPS2 Antibody in Western Blot (WB)
            PRPS2 Antibody in Immunocytochemistry (ICC/IF)
            PRPS2 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
            PRPS2 Antibody in Western Blot (WB)

            Product Details

            PA5-42007

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            0.2-1 µg/mL
            View 1 publication 1 publication

            Immunohistochemistry (IHC)

            -
            View 1 publication 1 publication

            Immunohistochemistry (Paraffin) (IHC (P))

            4-8 µg/mL
            View 1 publication 1 publication

            Immunocytochemistry (ICC/IF)

            -
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Human

            Published species

            Mouse

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Synthetic peptide directed towards the middle region of human PRPS2

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.5 mg/mL

            Purification

            Affinity Chromatography

            Storage buffer

            PBS with 2% sucrose

            Contains

            0.09% sodium azide

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2609479

            Product Specific Information

            Peptide sequence: VSPDAGGAKR VTSIADRLNV EFALIHKERK KANEVDRMVL VGDVKDRVAI

            Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%

            Target Information

            The Phosphoribosyl Pyrophosphate Synthetase 2 (PRPS2) gene plays a critical role in cellular metabolism by encoding an enzyme that conducts a key reaction linking central carbon metabolism and nucleotide production, essential for various biological functions. Structurally, PRPS2 shares a high sequence identity with PRPS1, making it less sensitive to allosteric inhibition and particularly significant in certain oncology contexts due to its upregulation at the translational level in some cancers. The enzyme encoded by PRPS2 is pivotal in coupling protein and nucleotide biosynthesis, aided by a unique cis-regulatory element in its 5' UTR, which is influenced by oncogenic pathways. This gene manifests a tissue-specific expression profile, predominantly in proliferative tissues such as the thymus, lung, and small intestine, highlighting its significance in cell growth and proliferation. The evolutionary origins of PRPS2, along with its functional diversification into multiple PRPS genes in eukaryotes, underscore its indispensable role in cellular physiology and pathology.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: Phosphoribosyl pyrophosphate synthase II; PPRibP; PPRibP synthetase; PRS-II; ribose-phosphate diphosphokinase 2; Ribose-phosphate pyrophosphokinase 2; testicular secretory protein Li 41

            View more View less

            Gene Aliases: PRPS2; PRSII

            View more View less

            UniProt ID: (Human) P11908

            View more View less

            Entrez Gene ID: (Human) 5634

            View more View less

            Function(s)

            magnesium ion binding ribose phosphate diphosphokinase activity protein binding ATP binding kinase activity identical protein binding protein homodimerization activity nucleotide kinase

            Process(es)

            5-phosphoribose 1-diphosphate biosynthetic process nucleobase-containing compound metabolic process nucleoside metabolic process ribonucleoside monophosphate biosynthetic process nucleotide biosynthetic process phosphorylation
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mgmkt:80/100.66.75.22:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0