Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: AKSPDAAKRA QSYAERLRLG LAVIHGEAQC TELDMDDGRH SPPMVKNATV HPGLELPLMM AKEKPPITVV GDVGGRIAII VDDIIDDVES FV
Seems to play a negative regulatory role in 5-phosphoribose 1-diphosphate synthesis. Target protein binds catalytic subunits of the ribose-phosphate diphosphokinase enzyme complex; may act as a negative regulator of enzyme activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 39 kDa phosphoribosypyrophosphate synthase-associated protein; kpra; PAP39; Phosphoribosyl pyrophosphate synthase-associated protein 1; phosphoribosyl pyrophosphate synthetase-associated protein 1; PRPP synthase-associated protein 1
Gene Aliases: PAP39; PRPSAP1
UniProt ID: (Human) Q14558
Entrez Gene ID: (Human) 5635
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support