Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VTPPELETKM NITKGGLVLF SANSNSSCME LSKKIAERLG VEMGKVQVYQ EPNRETRVQI QES
Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
The enzyme phosphoribosylpyrophosphate synthetase (PRS) catalyzes the formation of phosphoribosylpyrophosphate which is a substrate for synthesis of purine and pyrimidine nucleotides, histidine, tryptophan and NAD. PRS exists as a complex with two catalytic subunits and two associated subunits. This gene encodes a non-catalytic associated subunit of PRS.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 41 kDa phosphoribosypyrophosphate synthetase-associated protein; PAP41; Phosphoribosyl pyrophosphate synthase-associated protein 2; phosphoribosyl pyrophosphate synthetase-associated protein 2; PRPP synthase-associated protein 2
Gene Aliases: PAP41; PRPSAP2
UniProt ID: (Human) O60256
Entrez Gene ID: (Human) 5636
Molecular Function: nucleotide kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support