Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Peptide Sequence: QSTDNDVNYE DFTFTIPDVE DSSQRPDQGP QRPPPEGLLP RPPGDSGNQD DGPQQRPPKP GGHHRHPPPP PFQNQQRPPQ RGHRQLSLPR FPSVSLQEAS
Lacrimal proline rich protein is a member of the proline-rich protein family which lacks a conserved repetitive domain. It may have a role in protective functions in the eye. Two alternatively spliced transcript variants that encode different proteins have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lacrimal proline-rich protein; Nasopharyngeal carcinoma-associated proline-rich protein 4; proline-rich polypeptide 4; Proline-rich protein 4
Gene Aliases: LPRP; PROL4; PRR4
UniProt ID: (Human) Q16378
Entrez Gene ID: (Human) 11272
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support