Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRRSRSSAAA KLRGQKRSGA SGASAAPAAS AAAALAPSAT RTRRSASQAG SKSQAVEKPP SEKPRLRRSS PRAQEEGPGE PPPPELALLP PPPPPPPTPA TPTSSASNLD LGEQRERWET FQKRQKLTSE GAAKLLLDTF EYQGLVKHTG GCHCGAVRFE VWASADLHIF DCNCSICKKK QNRHFIVPAS RFKLLKGAEH ITTYTFNTHK AQHTFCKRCG VQSFYTPRSN PGGFGIAPHC LDEGTVRSMV TEEFNGSDWE KAMKEHKTIK NMSKE
CENPV orthologs were detected in a range of species, including plants, but the proline-rich region was detected only in primates, rodents, birds, and amphibians, and long proline stretches were found only in primates. Northern blot analysis showed ubiquitous Cenpv expression in mouse tissues, with highest expression in testis, followed by small intestine, brain, and kidney. Immunofluorescence microscopy of mouse fibroblasts revealed colocalization of Cenpv with stabilized cytoplasmic microtubules. Western blot analysis of Ramos B cells detectedCENPV at an apparent molecular mass of 40 kD.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CENP-V; Centromere protein V; Nuclear protein p30; proline rich 6; Proline-rich protein 6
Gene Aliases: 3110013H01Rik; CENP-V; CENPV; p30; PRR6
UniProt ID: (Human) Q7Z7K6
Entrez Gene ID: (Human) 201161
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support