Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: QLVQPGPHAM YARVRQVESN PLYQGTASSA DVALVELEAP VPFTNYILPV CLPDPSVIFE TGMNCWVTGW GSPSEEDLLP EPRILQKLAV PIIDTPKCNL LYSKDTEFGY
Pancreasin is a pancreatic tryptic serine peptidase that cleaves peptides after an arginine residue., Sep 2003].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: channel-activating protease 2; EC 3.4.21; EC 3.4.21.4; EC 3.4.21.43; Marapsin; MPNMarapsin; Pancreasin; Serine protease 27
Gene Aliases: CAPH2; MPN; PRSS27; UNQ1884/PRO4327
UniProt ID: (Human) Q9BQR3
Entrez Gene ID: (Human) 83886
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support