Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MEDYLQGCRA ALQESRPLHV VLGNEACDLD STVSALALAF YLAKTTEAEE VFVPVLNIKR SELPLRGDIV FFLQKVHIPE SILIFRDEID LHALYQAGQL TLILVDHHIL SKSDTALEEA VAEVLDHRPI EPKHCPPCHV SVELVGSCAT LVTERILQGA PEILDRQTAA LLHGTIILDC VNMDLKIGKA TPKDSKYVEK LEALFPDLPK RNDIFDSLQK AKFDVSGLTT EQMLRKDQKT IYRQGVKVAI SAIYMDLEAF LQRSNLLADL HAFCQAHSYD VLVAMTIFFN THNEPVRQLA IFCPHVALQT TICEVLERSH SPPLKLTPAS STHPNLHAYL QGNTQVSRKK LLPLLQEALS AYFDSMKIPS GQPETADVSR EQVDKELDRA SNSLISGLSQ DEEDPPLPPT PMNSLVDECP LDQGLPKLSA EAVFEKCSQI SLSQSTTASL SKK
Phosphodiesterase (PDE) that has higher activity toward cAMP than cGMP, as substrate. It plays a role in cell proliferation, is able to induce cell motility and acts as a negative regulator of NME1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DRES-17; Drosophila-related expressed sequence 17; Exopolyphosphatase PRUNE1; hPrune; HTcD37; Protein prune homolog 1
Gene Aliases: DRES-17; DRES17; H-PRUNE; HTCD37; PRUNE; PRUNE1
UniProt ID: (Human) Q86TP1
Entrez Gene ID: (Human) 58497
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support