Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Recommended positive controls: A549, HeLa, HepG2, HCT116.
Predicted reactivity: Human (99%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The human pregnancy-specific glycoproteins (PSGs) are a group of molecules that are mainly produced by the placental syncytiotrophoblasts during pregnancy. PSGs comprise a subgroup of the carcinoembryonic antigen (CEA) family, which belongs to the immunoglobulin superfamily. For additional general information about the PSG gene family, see PSG1 (MIM 176390).
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: pregnancy specific beta-1-glycoprotein 11; Pregnancy-specific beta-1 glycoprotein B; Pregnancy-specific beta-1-glycoprotein 11; pregnancy-specific beta-1-glycoprotein 11 C-R domain; Pregnancy-specific beta-1-glycoprotein 9; Pregnancy-specific glycoprotein 7; PS-beta-B; PS-beta-G-11; PS-beta-G-9; PS34; PSG7
Gene Aliases: PS34; PSBG-11; PSBG-9; PSG11; PSG9; PSGII
UniProt ID: (Human) Q00887
Entrez Gene ID: (Human) 5678
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support