Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSRGSSAGF DRHITIFSPE GRLYQVEYAF KAINQGGLTS VAVRGKDCAV IVTQKKVPDK LLDSSTVTHL FKITENIGCV MTGMTADSRS QVQRARYEAA NWKYKYGYEI PVDMLCKRIA DISQVYTQNA EMRPLGCCMI LIGIDEEQGP QVYKCDPAGY YCGFKATAAG VKQTESTSFL EKKVKKKFDW TFEQTVETAI TCLSTVLSID FKPSEIEVGV VTVENPKFRI LTEAEIDAHL VALAERD (1-246 aa encoded by BC023659)
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit. A pseudogene has been identified on the Y chromosome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 27 kDa prosomal protein; alpha-1; Macropain iota chain; macropain subunit iota; MGC22756; MGC2333; MGC23846; Multicatalytic endopeptidase complex iota chain; PROS 27; PROS-27; prosomal P27K protein; proteasome (prosome, macropain) subunit, alpha type, 6; proteasome alpha 6 subunit; Proteasome iota chain; Proteasome subunit alpha type-6; Proteasome subunit alpha-1; proteasome subunit iota; PSA6; PSMA 6; testicular secretory protein Li 44
Gene Aliases: IOTA; p27K; PROS27; PSMA6
UniProt ID: (Human) P60900, (Mouse) Q9QUM9, (Rat) P60901
Entrez Gene ID: (Human) 5687, (Mouse) 26443, (Rat) 29673
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support