Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: GFDDDGISRL YQTDPSGTYH AWKANAIGRS AKTVREFLEK NYTEDAIASD SEAIKLAIKA LLEVVQSGGR NIELAIIRRN QPLKMFSAKE VELYVTEIEK
Component of the spermatoproteasome, a proteasome specifically found in testis that promotes acetylation-dependent degradation of histones, thereby participating actively to the exchange of histones during spermatogenesis. The proteasome is a protein complexe that degrades unneeded or damaged proteins by proteolysis, a chemical reaction that breaks peptide bonds. Required for 20S core proteasome assembly, essential for the degradation of meiotic proteins RAD51 and RPA1 at late prophase I and the progression of meiosis I during spermatogenesis. Localizes to the synaptonemal complex, a 'zipper'-like structure that holds homologous chromosome pairs in synapsis during meiotic prophase I. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha4s; proteasome (prosome, macropain) subunit, alpha type, 8; Proteasome alpha 4 subunit; proteasome alpha 8 subunit; proteasome subunit alpha type 7-like protein variant 1; proteasome subunit alpha type-7-like; Proteasome subunit alpha type-8; Proteasome subunit alpha-type 7-like; Proteasome subunit alpha-type 8
Gene Aliases: 2410072D24Rik; PSMA7L; PSMA8; RGD1311659
UniProt ID: (Human) Q8TAA3, (Mouse) Q9CWH6
Entrez Gene ID: (Human) 143471, (Mouse) 73677, (Rat) 364814
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support