Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Positive control: Hela cell lysate, K562 cell lysate, 293T cell lysate, Jurkat cell lysate, Hela, Human skin tissue, Human breast tissue.
Predicted band size: 23 kDa
Subcellular Location: Cytoplasm, Nucleus, Proteasome.
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit. Pseudogenes have been identified on chromosomes 2 and 12.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta-3; MGC4147; proteasome (prosome, macropain) subunit, beta type, 3; Proteasome chain 13; Proteasome component C10-II; Proteasome subunit beta type-3; Proteasome subunit beta-3; Proteasome theta chain
Gene Aliases: HC10-II; PSMB3
UniProt ID: (Human) P49720
Entrez Gene ID: (Human) 5691
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support