Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EFIRNQEQM KPLEEKQEEE RSKVDDLRGT PMSVGTLEEI IDDNRAIVST SVGSEHYVSI LSFVDKDLLE PGCSVLLNHK VHAVIGVLMD DTDPLVTVMK VEKAPQETYA DIGGLDNQIQ EIKESVELPL THPEYYEEMG IKPPKGVILY GPPGTGKTLL AKAVANQTSA TFLRVVGSEL IQKYLGDGPK LVRELFRVAE EHAPSIVFID EIDAIGTKRY DSNSGGEREI QRTMLELLNQ LDGFDSRGDV KVIMATNRIE TLDPALIRPG RIDRKIEFPL PDEKTKKRIF QIHTSRMTLA DDVTLDDLIM AKDDLSGADI KAICTEAGLM ALRERRMKVT NEDFKKSKEN VLYKKQEGTP EGLYL (77-440 aa encoded by B C016368)
The 26S protease is involved in the ATP-dependent degradation of ubiquitinated proteins. The regulatory (or ATPase) complex confers ATP dependency and substrate specificity to the 26S complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 26S protease regulatory subunit 4; 26S proteasome AAA-ATPase subunit RPT2; 26S proteasome regulatory subunit 4; MGC24583; MGC8541; P26s4; peptidase (prosome, macropain) 26S subunit, ATPase 1; protease (prosome, macropain) 26S subunit, ATPase 1; proteasome (prosome, macropain) 26S subunit, ATPase, 1; proteasome 26S ATPase subunit 1; Proteasome 26S subunit ATPase 1; proteasome 26S subunit, ATPase, 1; prs4
Gene Aliases: AI325227; P26S4; p56; PSMC1; rpt2; Rpt2/S4; S4
UniProt ID: (Human) P62191, (Rat) P62193, (Mouse) P62192
Entrez Gene ID: (Human) 5700, (Rat) 117263, (Mouse) 19179
Molecular Function:
protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support