Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Positive control: MCF-7 cell lysate, Hela cell lysate, 293T cell lysate, mouse heart tissue lysate, mouse brain tissue lysate, rat testis tissue lysate, rat brain tissue, human colon tissue, human skin tissue, human kidney tissue.
Predicted band size: 46 kDa
Subcellular Location: Cytoplasm, Nucleus.
Human SUG1 (also known as p45, Rpt6 and Cim3) is a component of the 19S regulatory subunit of the 26S proteosome complex. It is one of the AAA ATPas found in the 19S regulatory complex that is thought to participate in the unfolding of ubiquitinated proteins in an ATP-dependent manner. It has also been shown to directly interact with the nucleotide excision repair protein XPB.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 26S protease regulatory subunit 8; 26S proteasome AAA-ATPase subunit RPT6; 26S proteasome regulatory subunit 8; for proteasomal ATPase (SUG1); mSUG1; MSUG1 protein; p45/SUG; peptidase (prosome, macropain) 26S subunit, ATPase 5; protease (prosome, macropain) 26S subunit, ATPase 5; proteasomal ATPase (SUG1); proteasome (prosome, macropain) 26S subunit, ATPase, 5; proteasome 26S ATPase subunit 5; Proteasome 26S subunit ATPase 5; proteasome p45/SUG; Proteasome subunit p45; Tat-binding protein homolog 10; testicular tissue protein Li 149; thy; Thyroid hormone receptor-interacting protein 1; thyroid receptor interactor 1; TRIP1
Gene Aliases: mSUG1; p45; p45/SUG; PSMC5; S8; SUG-1; SUG1; TBP10; TRIP1
UniProt ID: (Human) P62195, (Mouse) P62196, (Rat) P62198
Entrez Gene ID: (Human) 5705, (Mouse) 19184, (Rat) 81827
Molecular Function:
protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support