Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
This antibody is specific to PSMD4.
Immunogen sequence: MVLESTMVC VDNSEYMRNG DFLPTRLQAQ QDAVNIVCHS KTRSNPENNV GLITLANDCE VLTTLTPDTG RILSKLHTVQ PKGKITFCTG IRVAHLALKH RQGKNHKMRI IAFVGSPVED NEKDLVKLAK RLKKEKVNVD IINFGEEEVN TEKLTAFVNT LNGKDGTGSH LVTVPPGPSL ADALISSPIL AGEGGAMLGL GASDFEFGVD PSADPELALA LRVSMEEQRQ RQEEEARRAA AASAAEAGIA TTGTEDSDDA LLKMTISQQE FGRTGLPDLS SMTEEEQIAY AMQMSLQGAE FGQAESADID ASSAMDTSEP AKEEDDYDVM QDPEFLQSVL ENLPGVDPNN EAIRNAMGSL ASQATKDGKK DKKEEDKK (1-377 aa encoded by BC002365)
This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit functions as guanine nucleotide exchange factor. It is reported that this subunit interacts with HIV-1 Tat, and thus it represses the translation of host-cell, but not HIV-1, mRNAs. Several alternatively spliced transcript variants encoding multiple isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 26S proteasome non-ATPase regulatory subunit 4; 26S proteasome regulatory subunit RPN10; 26S proteasome regulatory subunit S5A; AF; angiocidin; anti secretory factor 1; antisecretory factor; Antisecretory factor 1; Multiubiquitin chain-binding protein; multiubiquitin-chain-binding protein; proteasome (prosome, macropain) 26S subunit, non-ATPase, 4; RP11-126K1.1; RPN10 homolog; S5a/antisecretory factor protein
Gene Aliases: AF; AF-1; Af1; angiocidin; ASF; MCB1; PSMD4; pUB-R5; Rpn10; S5A
UniProt ID: (Human) P55036, (Mouse) O35226
Entrez Gene ID: (Human) 5710, (Mouse) 19185, (Rat) 83499
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support