Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MVVTANGNDS KKFKGEDKMD GAPSRVLHIR KLPGEVTETE VIALGLPFGK VTNILMLKGK NQAFLELATE EAAITMVNYY SAVTPHLRNQ PIYIQYSNHK ELKTDNTLNQ RAQAVLQAVT AVQTANTPLS GTTVSESAVT P
PTBP2 encodes a protein that binds to the intronic cluster of RNA regulatory elements, downstream control sequence (DCS). It is implicated in controlling the assembly of other splicing-regulatory proteins. This protein is very similar to the polypyrimidine tract binding protein but it is expressed primarily in the brain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: br n nPTB5; neural polypyrimidine tract binding protein; Neural polypyrimidine tract-binding protein; Neurally-enriched homolog of PTB; nPTB5; nPTB6; nPTB7; nPTB8; Polypyrimidine tract-binding protein 2; PTB-like protein
Gene Aliases: brPTB; NPTB; PTB; PTBLP; PTBP2
UniProt ID: (Human) Q9UKA9
Entrez Gene ID: (Human) 58155
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support