Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Mutations of the human Patched tumor suppressor gene (PTCH) have been identified in individuals with the nevoid basal cell carcinoma syndrome (NBCCS) as well as in sporadic basal cell carcinomas and medulloblastomas. PTCH2 has a frameshift mutation resulting in a truncated PTCH2 protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: patched homolog 2; Protein patched homolog 2; PTC2; RP11-269F19.8
Gene Aliases: PTC2; PTCH2; UNQ560/PRO1121/PRO57079
UniProt ID: (Human) Q9Y6C5
Entrez Gene ID: (Human) 8643
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support