Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSSLSGKVQ TVLGLVEPSK LGRTLTHEHL AMTFDCCYCP PPPCQEAISK EPIVMKNLYW IQKNAYSHKE NLQLNQETEA IKEELLYFKA NGGGALVENT TTGISRDTQT LKRLAEETGV HIISGAGFYV DATHSSETRA MSVEQLTDVL MNEILHGADG TSIKCGIIGE IGCSWPLTES ERKVLQATAH AQAQLGCPVI IHPGRSSRAP FQIIRILQEA GADISKTVMS HLDRTILDKK ELLEFAQLGC YLEYDLFGTE LLHYQLGPDI DMPDDNKRIR RVRLLVEEGC EDRILVAHDI HTKTRLMKYG GHGYSHILTN VVPKMLLRGI TENVLDKILI ENPKQWLTFK (1-349 aa encoded by BC015092)
PTER is a mammalian homolog to bacterial phosphotriesterases, enzymes that hydrolyze phosphotriester-containing organophosphate pesticides. It is expressed primarily in the proximal renal tubules and the gene has been localized in humans to chromosomal band 10p12 by in situ hybridization. PTER, in addition to FTO, MC4R, and NPC1 has recently been shown to be a new risk loci for early-onset and morbid adult obesity in European populations. At least two isoforms of PTER are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPHRP; Parathion hydrolase-related protein; Phosphotriesterase-related protein; resiniferatoxin-binding phosphotriesterase-related protein; resiniferatoxin-binding, phosphotriesterase-related; resiniferatoxin-binding, phosphotriesterase-related protein; Resiniferotoxin-binding phosphotriesterase-related protein
Gene Aliases: AI790318; HPHRP; Mpr56-1; PTER; RPR-1; Rpr1
UniProt ID: (Human) Q96BW5, (Rat) Q63530, (Mouse) Q60866
Entrez Gene ID: (Human) 9317, (Rat) 63852, (Mouse) 19212
Molecular Function:
esterase
hydrolase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support