Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: RKAVFQRLKL WVCCLCLGPA HGDSQTPLSQ LASGRRDPRA PSAPVGKEGS CVPLSAWGEG QVEPLPPTQQ SSGSAVGTSS KAEASVACSL C
Receptor for prostacyclin (prostaglandin I2 or PGI2). The activity of this receptor is mediated by G(s) proteins which activate adenylate cyclase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IP receptor; PGI receptor; PGI2 receptor; Prostacyclin receptor; Prostaglandin I2 receptor; Prostanoid IP receptor
Gene Aliases: IP; PRIPR; PTGIR
UniProt ID: (Human) P43119
Entrez Gene ID: (Human) 5739
Molecular Function: G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support