Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MVSRDQAHLG PKYVGLWDFK SRTDEELSFR AGDVFHVARK EEQWWWATLL DEAGGAVAQG YVPHNYLAER ETVESEPWFF GCISRSEAVR RLQAEGNATG AFLIRVSEKP SADYVLSVRD TQAVRHYKIW RRAGGRLHLN EAVSFLSLPE LVNYHRAQSL SHGLRLAAPC RKHEPEPLPH WDDWERPREE FTLCRKLGSG YFGEVFEGLW KDRVQVAIKV ISRDNLLHQQ MLQSEIQAMK KLRHKHILAL YAVVSVGDPV YIITELMAKG SLLELLRDSD EKVLPVSELL DIAWQVAEGM CYLESQNYIH RDLAARNILV GENTLCKVGD FGLARLIKED VYLSHDHNIP YKWTAPEALS RGHYSTKSDV WSFGILLHEM FSRGQVPYPG MSNHEAFLRV DAGYRMPCPL ECPPSVHKLM LTCWCRDPEQ RPCFKALRER LSSFTSYENP T
Breast tumor kinase (Brk) is a non-receptor protein tyrosine kinase expressed in over 60% of breast carcinoma tissue samples and breast tumor cell lines. Brk is 80% identity to Sik (Src-related intestinal kinase) and exhibits the features of a novel non-receptor tyrosine kinase, including amino terminal SH3 and SH2 domains. Low levels of Brk transcripts have been identified in some human breast tumor cell lines but not in normal breast tissue.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ALT-PTK6; Breast tumor kinase; deltam5; FLJ42088; Protein-tyrosine kinase 6; protein-tyrosine kinase BRK; PTK6 protein tyrosine kinase 6; Tyrosine-protein kinase BRK
Gene Aliases: BRK; PTK6
UniProt ID: (Human) Q13882
Entrez Gene ID: (Human) 5753
Molecular Function: protein-binding activity modulator scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support