Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
This antibody is predicted to react with mouse and rat based on sequence homology.
PRL-2, also known as Protein tyrosine phosphatase 4a2 (Ptp4a2) is a unique nuclear protein tyrosine phosphatase (PTP) that plays a central role in regulating diverse cellular processes. PRL-1 is induced in mitogen-stimulated cells and regenerating liver. PRL-2 exhibits 87% identity to PRL-1 in their amino acid sequences. All mouse PRL proteins contain a C-terminal consensus sequence for prenylation. All PRL proteins bear significant sequence homology to Cdc14p and the tumor suppressor PTEN/MMAC1. PRL-2 is preferentially expressed in skeletal muscle. PRL-2 is also expressed at lower levels in other tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HU-PP-1; OV-1; phosphatase of regenerating liver 2; PRL-2; protein tyrosine phosphatase IVA; protein tyrosine phosphatase IVA2; Protein tyrosine phosphatase type IVA 2; Protein-tyrosine phosphatase 4a2; Protein-tyrosine phosphatase of regenerating liver 2; PTP(CAAXII); tyrosine-phosphatase
Gene Aliases: BM-008; HH13; HH7-2; HU-PP-1; OV-1; PRL-2; PRL2; ptp-IV1a; ptp-IV1b; PTP4A; PTP4A2; PTPCAAX2
UniProt ID: (Human) Q12974
Entrez Gene ID: (Human) 8073
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support