Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAATALLEA GLARVLFYPT LLYTLFRGKV PGRAHRDWYH RIDPTVLLGA LPLRSLTRQL VQDENVRGVI TMNEEYETRF LCNSSQEWKR LGVEQLRLST VDMTGIPTLD NLQKGVQFAL KYQSLGQCVY VHCKAGRSRS ATMVAAYLIQ VHKWSPEEAV RAIAKIRSYI HIRPGQLDVL KEFHKQITAR ATKDGTFVIS KT (1-201 aa encoded by BC020242)
Lipid phosphatase which dephosphorylates phosphatidylglycerophosphate (PGP) to phosphatidylglycerol (PG). PGP is an essential intermediate in the biosynthetic pathway of cardiolipin, a mitochondrial-specific phospholipid regulating the membrane integrity and activities of the organelle. Has also been shown to display phosphatase activity toward phosphoprotein substrates, specifically mediates dephosphorylation of mitochondrial proteins, thereby playing an essential role in ATP production. Has probably a preference for proteins phosphorylated on Ser and/or Thr residues compared to proteins phosphorylated on Tyr residues. Probably involved in regulation of insulin secretion in pancreatic beta cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NB4 apoptosis/differentiation related protein; Phosphatidylglycerophosphatase and protein-tyrosine phosphatase 1; Phosphoinositide lipid phosphatase; Protein-tyrosine phosphatase mitochondrial 1; PTEN-like phosphatase
Gene Aliases: 1110001D10Rik; 2810004N20Rik; DUSP23; MOSP; PLIP; PNAS-129; PTPMT1
UniProt ID: (Human) Q8WUK0, (Mouse) Q66GT5
Entrez Gene ID: (Human) 114971, (Mouse) 66461, (Rat) 29390
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support