Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTSRRWFHP NITGVEAENL LLTRGVDGSF LARPSKSNPG DFTLSVRRNG AVTHIKIQNT GDYYDLYGGE KFATLAELVQ YYMEHHGQLK EKNGDVIELK YPLNCADPTS ERWFHGHLSG KEAEKLLTEK GKHGSFLVRE SQSHPGDFVL SVRTGDDKGE SNDGKSKVTH VMIRCQELKY DVGGGERFDS LTDLVEHYKK NPMVETLGTV LQLKQPLNTT RINAAEIESR VRELSKLAET TDKVKQGFWE EFETLQQQEC KLLYSRKEGQ RQENKNKNRY KNILPFDHTR VVLHDGDPNE PVSDYINANI IMPEFETKCN NSKPKKSYIA TQGCLQNTVN DFWRMVFQEN SRVIVMTT (1-357 aa encoded by BC008692)
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains two tandem Src homology-2 domains, which function as phospho-tyrosine binding domains and mediate the interaction of this PTP with its substrates. This PTP is widely expressed in most tissues and plays a regulatory role in various cell signaling events that are important for a diversity of cell functions, such as mitogenic activation, metabolic control, transcription regulation, and cell migration. Mutations in this gene are a cause of Noonan syndrome as well as acute myeloid leukemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: encodes catalytic domain; HCP; HGNC:9644; MGC14433; Noonan syndrome 1; null; OTTHUMP00000166108; protein tyrosine phosphatase SH-PTP2; Protein-tyrosine phosphatase 1D; Protein-tyrosine phosphatase 2C; Protein-tyrosine phosphatase SYP; PTP-1D; PTP-2C; PTP1C; SH-PTP2; SH-PTP2 protein tyrosine phosphatase non-receptor type 11; SH-PTP2 protein tyrosine phosphatase, non-receptor type 11; SH-PTP3; SH2 domain-containing protein tyrosine phosphatase-2; SHP-2; Tyrosine-protein phosphatase non-receptor type 11
Gene Aliases: 2700084A17Rik; AW536184; BPTP3; CFC; JMML; METCDS; NS1; PTP-1D; PTP1D; PTP2C; PTPN11; SAP-2; SH-PTP2; SH-PTP3; SHP-2; SHP2; SHPTP2; Syp
UniProt ID: (Human) Q06124, (Mouse) P35235, (Rat) P41499
Entrez Gene ID: (Human) 5781, (Mouse) 19247, (Rat) 25622
Molecular Function: protein phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support