Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).^M
^M
Immunogen sequence: PESFVLASEH NTPVRSEWSE LQSQERSEQK KSEGLITSEN EKCDHPAGGI HYEMCIECPP TFSDKREQIS ENPTEATDIG FGNRCGKPKG PRDPPSEW
PTP PEST is an 88 kDa cytosolic protein tyrosine phosphatase which is ubiquitiously expressed in mammalian tissues. PTP PEST exhibits a high specific activity when assayed in vitro with artificial tyrosine-phosphorylated substrates. PTP PEST is subject to regulation via phosphorylation of ser 39 and is catalyzed by both protein kinase C and protein kinase A.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: pest; Protein-tyrosine phosphatase G1; PTP-PEST; PTPG1; tcag7.1075; Tyrosine-protein phosphatase non-receptor type 12
Gene Aliases: PTP-PEST; PTPG1; PTPN12
UniProt ID: (Human) Q05209
Entrez Gene ID: (Human) 5782
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support