Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 16
Antibody detects endogenous levels of total PTPN23.
Plays a role in sorting of endocytic ubiquitinated cargos into multivesicular bodies (MVBs) via its interaction with the ESCRT-I complex (endosomal sorting complex required for transport I), and possibly also other ESCRT complexes. May act as a negative regulator of Ras-mediated mitogenic activity. Plays a role in ciliogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HD-PTP; His domain-containing protein tyrosine phosphatase; Protein tyrosine phosphatase TD14; PTP-TD14; Tyrosine-protein phosphatase non-receptor type 23
Gene Aliases: AI462446; HD-PTP; HDPTP; KIAA1471; PTP-TD14; PTPN23
UniProt ID: (Human) Q9H3S7, (Mouse) Q6PB44
Entrez Gene ID: (Human) 25930, (Mouse) 104831, (Rat) 117552
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support