Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 4
Immunogen sequence: MTSRFRLPAG RTYNVRASEL ARDRQHTEVV CNILLLDNTV QAFKVNKHDQ GQVLLDVVFK HLDLTEQDYF GLQLADDSTD NPRWLDPNKP IRKQLKRGSP YSLNFRVKFF VSDPNKLQEE YTRYQYFLQI KQDILTGRLP CPSNTAALLA SFAVQSELGD YDQSENLSGY LSDYSFIPNQ PQDFEKEIAK LHQQHIGLSP AEAEFNYLNT ARTLELYGVE FHYARDQSNN EIMIGVMSGG
The protein encoded by this gene is a member of the protein tyrosine phosphatase family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This protein contains a C-terminal PTP domain and an N-terminal domain homologous to the band 4.1 superfamily of cytoskeletal-associated proteins. This PTP has been shown to interact with glutamate receptor delta 2 and epsilon subunits, and is thought to play a role in signaling downstream of the glutamate receptors through tyrosine dephosphorylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MEG; megakaryocyte phosphatase; megakaryocyte protein-tyrosine phosphatase; protein tyrosine phosphatase MEG1; protein tyrosine phosphatase, non-receptor type 4 (megakaryocyte); Protein-tyrosine phosphatase MEG1; PTP PTPMEG1; PTPase-MEG1; Testis-enriched protein tyrosine phosphatase; Tyrosine-protein phosphatase non-receptor type 4
Gene Aliases: hPTP-MEG; MEG; Ptn4; PTPMEG; PTPMEG1; PTPN4; TEP; TEP/mPTPMEG
UniProt ID: (Human) P29074, (Mouse) Q9WU22
Entrez Gene ID: (Human) 5775, (Mouse) 19258, (Rat) 246116
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support