Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Possible cell adhesion receptor. It possesses an intrinsic protein tyrosine phosphatase activity (PTPase).The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ43335; FLJ45062; FLJ45567; LAR; LAR protein; LCA homolog; LCA-homolog; leukocyte antigen-related (LAR) PTP receptor; leukocyte antigen-related tyrosine phosphatase; Leukocyte common antigen related; protein tyrosine phosphatase, receptor type, F polypeptide; PTPRF protein; receptor-linked protein-tyrosine phosphatase LAR; Receptor-type tyrosine-protein phosphatase F
Gene Aliases: BNAH2; LAR; PTPRF
UniProt ID: (Human) P10586
Entrez Gene ID: (Human) 5792
Molecular Function: protein phosphatase protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support