Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRRAVCFPAL CLLLNLHAAG CFSGNNDHFL AINQKKSGKP VFIYKHSQDI EKSLDIAPQK IYRHSYHSSS EAQVSKRHQI VNSAFPRPAY DPSLNLLAMD GQDLEVENLP IPAANVIVVT LQMDVNKLNI TLLRIFRQGV AAALGLLPQQ VHINRLIGKK NSIELFVSPI NRKTGISDAL PSEEVLRSLN INVLHQSLSQ FGITEVSPEK NVLQGQHEAD KIWSKEGFYA VVIFLSIFVI IVTCLMILYR LKERFQLSLR QDKEKNQEIH LSPITLQPAL SEAKTVHSMV QPEQAPKVLN VVVDPQGRGA PEIRATTATS VCPSPFKMKP IGLQERRGSN VSLTLDMSSL GNIEPFVSIP TPREKVAMEY LQSASRILTR SQLRDVVASS HLLQSEFMEI PMNFVDPKEI DIPRHGTKNR YKTILPNPLS RVCLRPKNVT DSLSTYINAN YIRGYSGKEK AFIATQGPMI NTVDDFWQMV WQEDSPVIVM ITKLKEKNEK CVLYWPEKRG IYGKVEVLVI SVNECDNYTI RNLVLKQGSH TQHVKHYWYT SWPDHKTPDS AQPLLQLMLD VEEDRLASQG RGPVVVHCSA GIGRTGCFIA TSIGCQQLKE EGVVDALSIV CQLRMDRGGM VQTSEQYEFV HHALCLYESR LSAETVQ
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and a single intracellular catalytic domains, and thus represents a receptor-type PTP. The similar gene predominately expressed in mouse brain was found to associate with, and thus regulate the activity and cellular localization of MAP kinases. The rat counterpart of this gene was reported to be regulated by the nerve growth factor, which suggested the function of this gene in neuronal growth and differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ch-1 PTPase; Ch-1PTPase; ECPPT; NC-PTPCOM1; protein tyrosine phosphatase Cr1PTPase; protein tyrosine phosphatase, receptor type, R; protein-tyrosine phosphatase NC-PTPCOM1; Protein-tyrosine phosphatase PCPTP1; R-PTP-R; Receptor-type tyrosine-protein phosphatase R
Gene Aliases: EC-PTP; ECPTP; PCPTP1; PTP-SL; PTPBR7; PTPRQ; PTPRR
UniProt ID: (Human) Q15256
Entrez Gene ID: (Human) 5801
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support