Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene is a member of the protein tyrosine phosphatase family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and two tandem intracellular catalytic domains, and thus represents a receptor-type PTP. The extracellular region contains a meprin-A5 antigen-PTP domain, Ig-like and fibronectin type III-like repeats. The protein domain structure and the expression pattern of the mouse counterpart of this PTP suggest its roles in both signal transduction and cellular adhesion in the central nervous system. Two alternatively spliced transcript variants of this gene, which encode distinct proteins, have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: protein tyrosine phosphatase, receptor type, T; R-PTP-T; receptor protein tyrosine phosphatase; receptor protein tyrosine phosphatase-rho; Receptor-type tyrosine-protein phosphatase rho; Receptor-type tyrosine-protein phosphatase T; RP5-1121H13.2; RPTP-rho; RPTPmam4
Gene Aliases: KIAA0283; mKIAA0283; mRPTPrho; PTPRT; R-PTP-T; RPTP-rfo; RPTP-rho; RPTPmam4; RPTPrho
UniProt ID: (Human) O14522, (Mouse) Q99M80
Entrez Gene ID: (Human) 11122, (Mouse) 19281
Molecular Function:
extracellular matrix glycoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support