Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is human spleen tissue lysate.
PA5-34522 can be used with blocking peptide PEP-1565.
The polymerase I and transcript release factor (PTRF) enables the dissociation of paused ternary polymerase I transcription complexes from the 3' end of pre-rRNA transcripts, regulating rRNA transcription by promoting the dissociation of transcription complexes and the re-initiation of polymerase I on nascent rRNA transcripts. PTRF also localizes to caveolae at the plasma membrane and is thought to play a critical role in the formation of caveolae and the stabilization of caveolins, translocating from caveolae to the cytoplasm after insulin stimulation. PTRF is also thought to modify lipid metabolism and insulin-regulated gene expression. Mutations in this gene result in a disorder characterized by generalized lipodystrophy and muscular dystrophy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Caveolae-associated protein 1; Cavin-1; FLJ90031; Polymerase I and transcript release factor; PTRF; RNA polymerase I and transcript release factor; TTF-I interacting peptide 12
Gene Aliases: CAVIN; cavin-1; CAVIN1; CGL4; FKSG13; PTRF
UniProt ID: (Human) Q6NZI2
Entrez Gene ID: (Human) 284119
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support