Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product is supplied at 1 mL (100 µL/mL).
Application Notes: Western blot- A non-reduced sample treatment and SDS-Page was used. The band size is 400 kDa. Flow cytometry - This antibody stains the intra and extracellular domain of Pentraxin 3. For intracellular staining cells were permeabilized with buffer containing triton. Transfected cells were used as a positive control.
Pentraxins are a protein superfamily that is characterized by a cyclic multimeric structure. Ptx3, also known as tumor necrosis factor-stimulated gene sequence-14 (TSG14), is a secreted pattern-recognition receptor that has a non-redundant role in resistance to selected microbial agents. Ptx3 belongs to the family of "long pentraxins", which have C-terminal pentraxin domains and novel amino-terminal domains. Ptx3 binds selected pathogens, including Aspergillus fumigatus, Pseudomonas aeruginosa and Salmonella typhimurium. It is synthesized in IgA glomerulonephritis and activates mesangial cells. Secretion of Ptx3 in adipose cells can be induced by TNF. Ptx3 is also involved in amplification of inflammatory reactions and regulation of innate immunity. The human PTX3 gene maps to chromosome 3q25.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: pentaxin related; Pentaxin-related protein PTX3; pentraxin 3; Pentraxin-related protein PTX3; TSG-14; Tumor necrosis factor-inducible gene 14 protein
Gene Aliases: AI607804; Ptx3; TSG-14; Tsg14
UniProt ID: (Mouse) P48759
Entrez Gene ID: (Mouse) 19288
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support