Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Immunogen sequence: VQKLKAKLS SWCIPLTGAP NSDNTLATSG KECALCGERP TMPHTIGCEH IFCYFCAKSS FLFDVYFTCP KCGTEVHSLQ PLKSGIEMSE VNPL (213-305 aa encoded by B C000661)
This gene encodes an integral peroxisomal membrane protein required for peroxisome biogenesis. The protein is thought to be involved in peroxisomal matrix protein import. Mutations in this gene result in one form of Zellweger syndrome and infantile Refsum disease. Alternative splicing results in multiple transcript variants encoding the same protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 35 kDa peroxisomal membrane protein; PAF-1; peroxin 2; Peroxin-2; Peroxisomal membrane protein 3; peroxisomal membrane protein 3, 35 kDa; peroxisomal membrane protein 3, 35kDa; Peroxisome assembly factor 1; peroxisome assembly factor-1; Peroxisome biogenesis factor 2; RING finger protein 72; Zellweger syndrome homolog
Gene Aliases: D3Ertd138e; PAF-1; PAF1; PBD5A; PBD5B; PEX2; PMP3; PMP35; PXMP3; RNF72; ZWS3
UniProt ID: (Human) P28328, (Mouse) P55098
Entrez Gene ID: (Human) 5828, (Mouse) 19302
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support