Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Ankyrins are a family of adaptor proteins that mediate the attachment of integral membrane proteins to the spectrin-actin based membrane skeleton. Ankyrins have binding sites for the beta subunit of spectrin and at least 12 families of integral membrane proteins. This linkage is required to maintain the integrity of the plasma membranes and to anchor specific ion channels, ion exchangers and ion transporters in the plasma membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ANK-1; ANK-2; ANK-3; ank2; ankyrin 1, erythrocytic; ankyrin 1, erythroid; ankyrin 2, brain; ankyrin 3 (G); ankyrin 3, epithelial; ankyrin B; ankyrin, brain; Ankyrin-1; Ankyrin-2; ankyrin-2, nonerythrocytic; Ankyrin-3; Ankyrin-B; Ankyrin-G; Ankyrin-R; Brain ankyrin; brain-specific ankyrin-G; Erythrocyte ankyrin; Non-erythroid ankyrin; normoblastic anemia; rCG_43073
Gene Aliases: 2900054D09Rik; AI314020; AI835472; ANK; Ank-1; ANK-2; ANK-3; ANK1; ANK2; ANK3; ANKB; AnkG; Ankyrin-3; ANKYRIN-G; AW491075; brank-2; LQT4; MRT37; nb; pale; SPH1; SPH2
UniProt ID: (Human) P16157, (Human) Q01484, (Human) B4DIL1, (Mouse) Q02357, (Mouse) Q8C8R3, (Rat) O70511, (Mouse) G5E8K5
Entrez Gene ID: (Human) 286, (Human) 287, (Human) 288, (Mouse) 11733, (Rat) 306570, (Mouse) 109676, (Rat) 362036, (Rat) 361833, (Mouse) 11735
Molecular Function: scaffold/adaptor protein membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support