Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The antigen is homologous or shares a significant identity with many RAB molecules including RAB1, RAB2, RAB4, RAB9, RAB10, RAB11, RAB14, RAB26, RAB30, RAB35, RAB39, RAB43.
Specificity of this antibody: RAB.
Rab GTPases regulate many steps of membrane trafficking, including vesicle formation, vesicle movement along actin and tubulin networks, and membrane fusion by recruiting specific effector proteins to restricted membranes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 24KG; bA165P4.3 (member RAS oncogene family); F protein-binding protein 1; GTP binding protein Rab1a; GTP-binding protein; GTP-binding protein RAB10; GTP-binding protein RAB2B; GTP-binding protein RAY; GTP-binding protein YPT3; GTPase Rab14; HTLV-1 related endogenous sequence; RAB 11A, member oncogene family; Rab GTPase YPT1 homolog; Rab-11; Rab-39; Rab-9-like protein; rab-9A; Rab-9L; rab-related GTP-binding protein; RAB1, member RAS oncogene family; RAB11B, member of RAS oncogene family; RAB1B-like, member RAS oncogene family; RAB2, member RAS oncogene family; RAB39, member RAS oncogene family; RAB9, member RAS oncogene family; RAS family, member RAB2B; RAS oncogene family protein; RAS-associated protein RAB3A; ras-related GTP-binding protein; ras-related GTP-binding protein 4b; Ras-related oncogene protein; Ras-related protein Rab-10; Ras-related protein Rab-11A; Ras-related protein Rab-11B; Ras-related protein Rab-14; Ras-related protein Rab-1A; Ras-related protein Rab-1B; Ras-related protein Rab-1C; ras-related protein rab-1c (GTP-binding protein ray); Ras-related protein Rab-26; Ras-related protein Rab-2A; Ras-related protein Rab-2B; Ras-related protein Rab-30; Ras-related protein Rab-35; Ras-related protein Rab-39A; Ras-related protein Rab-39B; Ras-related protein Rab-3A; Ras-related protein Rab-41; Ras-related protein Rab-43; Ras-related protein Rab-4A; Ras-related protein Rab-4B; Ras-related protein Rab-9A; Ras-related protein Rab-9B; ras-related protein rab10; Ras-related protein RAB43; Ras-related small GTP binding protein 3A; Ras-related small GTP binding protein 4; ras-related YPT1 protein; SID 99; small GTP binding protein RAB14; small GTP binding protein RAB2A; small GTP binding protein RAB4B; small GTP-binding protein; YL8; YPT1-related protein
Gene Aliases: 0610030G24Rik; 1110011F09Rik; 1500012D09Rik; 1500031G17Rik; 1810048P08Rik; 2410064E05Rik; 2500004H21Rik; 2810475J17Rik; 4930528G15Rik; 5033421K01Rik; 6330580M05Rik; 9330148M11Rik; 9330195C02Rik; 9530019H02Rik; A230002G14; A730055L17Rik; A830020M03Rik; A830021G03Rik; Ac1075; Ac2-048; AI195561; AI314285; AI323892; AI649155; AI848268; AU040256; AW107754; AW490415; C230094F14Rik; C80220; D030017L14Rik; D530043M21Rik; FBP; Gtbp; H-ray; H-YPT3; HRES-1; HRES-1/RAB4; HRES1; LHX; mKIAA3012; MRX72; PP1596; Rab-1; RAB-14; Rab-9L; RAB1; RAB10; RAB11; RAB11A; RAB11B; RAB14; RAB1A; RAB1B; Rab1bl; RAB1C; Rab1r; RAB2; RAB26; RAB2A; RAB2B; RAB3; RAB30; RAB35; RAB39; RAB39A; RAB39B; RAB3A; RAB4; RAB41; RAB43; RAB4A; RAB4B; RAB9; RAB9A; RAB9B; RAB9L; RAY; RGD1562951; Rsb30; Sid6061p; Sid99; V46133; WSMN; YL8; YPT1; YPT3
UniProt ID: (Human) P61026, (Human) P62491, (Human) Q6FI42, (Human) P61106, (Human) P62820, (Human) Q9H0U4, (Human) Q9ULW5, (Human) P61019, (Human) Q8WUD1, (Human) Q15771, (Human) Q15286, (Human) Q14964, (Human) Q96DA2, (Human) P20336, (Human) Q86YS6, (Human) P20338, (Human) P61018, (Human) P51151, (Human) Q9NP90, (Mouse) P62821, (Mouse) P61027, (Rat) P62494, (Mouse) P62492, (Rat) O35509, (Mouse) P46638, (Mouse) Q91V41, (Rat) P61107, (Rat) P05711, (Mouse) Q9D1G1, (Mouse) Q504M8, (Rat) P51156, (Mouse) P53994, (Rat) P05712, (Mouse) P59279, (Mouse) Q923S9, (Mouse) Q6PHN9, (Rat) Q5U316, (Mouse) Q8BHD0, (Mouse) Q8BHC1, (Rat) P63012, (Mouse) P63011, (Mouse) Q8CG50, (Rat) Q53B90, (Rat) P05714, (Mouse) P56371, (Rat) P51146, (Mouse) Q91ZR1, (Mouse) Q9R0M6, (Rat) Q99P75, (Mouse) Q8BHH2
Entrez Gene ID: (Human) 10890, (Human) 8766, (Human) 9230, (Human) 51552, (Human) 5861, (Human) 81876, (Human) 25837, (Human) 5862, (Human) 84932, (Human) 27314, (Human) 11021, (Human) 54734, (Human) 116442, (Human) 5864, (Human) 339122, (Human) 5867, (Human) 53916, (Human) 9367, (Human) 51209, (Mouse) 19324, (Mouse) 19325, (Rat) 50993, (Rat) 81830, (Mouse) 53869, (Rat) 79434, (Mouse) 19326, (Mouse) 68365, (Rat) 94197, (Rat) 81754, (Rat) 100126191, (Mouse) 76308, (Mouse) 328778, (Rat) 171111, (Mouse) 59021, (Rat) 65158, (Rat) 305853, (Mouse) 76338, (Rat) 308821, (Mouse) 75985, (Mouse) 77407, (Rat) 288700, (Mouse) 270160, (Rat) 315668, (Mouse) 67790, (Rat) 25531, (Mouse) 19339, (Mouse) 69834, (Rat) 500249, (Rat) 25532, (Mouse) 19341, (Rat) 50866, (Mouse) 19342, (Mouse) 56382, (Rat) 84589, (Rat) 367915, (Mouse) 319642
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support