Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: VNCICVDWK GGSRTGYTQA SQNIRIVGAE VAYFVEFLQS AFGYSPSNVH VIGHSLGAHA AGEAGRRTNG TIGRITGLDP AEPCFQGTPE LVRLDPSDAK FVDVIHTDGA PIVPNLGFGM SQVVGHLDFF PNGGVEMPGC KKNILSQIVD IDGIWEGTRD FAACNHLRSY KYYTDSIVNP DGFAGFPCAS YNVFTANKCF PCPSGGCPQM GHYADRYPGK TNDVGQKFYL DTGDASNFAR WRYKVSVTLS GKKVTGHILV SLFGNKGNSK QYEIFKGTLK PDSTHSNEFD SDVDVGDLQM VKFIWYNNVI NPTLPRVGAS KIIVETNVGK QFNFCSPETV REEVLLTLTP C (116-465 aa encoded by BC014309)
This gene is a member of the lipase gene family. It encodes a carboxyl esterase that hydrolyzes insoluble, emulsified triglycerides, and is essential for the efficient digestion of dietary fats. This gene is expressed specifically in the pancreas.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Pancreatic triacylglycerol lipase; pancreatic triglyceride lipase; PL; RP11-33D13.1
Gene Aliases: 1810007A24Rik; PANLI; PL; PNLIP; PNLIPD; PTL
UniProt ID: (Human) P16233, (Rat) P27657, (Mouse) Q6P8U6
Entrez Gene ID: (Human) 5406, (Rat) 25702, (Mouse) 69060
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support