Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 4
Immunogen sequence: MSMTDLLNAE DIKKAVGAFS ATDSFDHKKF FQMVGLKKKS ADDVKKVFHM LDKDKSGFIE EDELGFILKG FSPDARDLSA KETKMLMAAG DKDGDGKIGV DEFSTLVAES
Parvalbumin (PV) is a calcium binding protein expressed in specific muscle fibers and fast-firing neurons. PV consists of a single, unbranched chain of linked amino acids and belongs to a larger group of EF hand proteins. Studies have demonstrated that parvalbumin acts in the decay of calcium in the contraction/ relaxation cycle of fast twitch muscles. This data has shown a positive correlation between the rate of relaxation and the concentration of parvalbumin. Parvalbumin is also expressed in a specific population of GABAergic interneurones which are thought to play a role in maintaining the balance between excitation and inhibition in the cortex as well as the hippocampus. In amyotrophic lateral sclerosis (ALS) patents, parvalbumin immunoreactivity is specifically absent from neuron populations lost early in ALS.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HGNC:9704; MGC116759; Parvalbumin (calcium binding protein); Parvalbumin alpha
Gene Aliases: D22S749; PALB1; Parv; PV; Pva; PVALB
UniProt ID: (Human) P20472, (Rat) P02625, (Mouse) P32848
Entrez Gene ID: (Human) 5816, (Rat) 25269, (Mouse) 19293
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support