Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: VLEGNPEVGS PRTSDLQHRG NKGSCVLSSP GEDAQPGEEP IKYGELIVLG
Sequence homology: Cow: 92%; Guinea Pig: 93%; Horse: 85%; Human: 100%; Mouse: 92%; Pig: 100%; Rabbit: 93%; Rat: 93%
Pellino 1 is the mammalian homologue of Pellino, a Drosophila Pelle-interacting protein. Pelle is a serine-threonine kinase through which Toll signaling can occur. Pellino 1 can activate c-Jun, Elk-1 and NF-kB, whereas Pellino 3 can only activate c-Jun and Elk-1(2).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase pellino homolog 3; MGC35521; pellino homolog 3; Pellino-3; protein pellino homolog 3
Gene Aliases: PELI3
UniProt ID: (Human) Q8TED5
Entrez Gene ID: (Human) 246330
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support