Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
PA1-1052 detects perilipin A and B from rat and 3T3-L1 cell extract.
PA1-1052 has been successfully used in Western blot procedures. By Western blot, this antibody detects ~62 and 46 kDa proteins representing perilipin A and B, respectively from 3T3-L1 cell extract
The PA1-1052 immunogen is a synthetic peptide corresponding to residues M(1) S M N K G P T L L D G D L P E Q(17) C of rat Perilipin A and B. This sequence is 82% conserved in humans. The PA1-1052 immunizing peptide (Cat. # PEP-170) is available for use in neutralization and control experiments.
Adipose tissue is an energy reserve in animals and is strictly regulated in nondomestic species. Adipose cells produce and secrete numerous physiologically important proteins, such as lipoprotein lipase (LPL), leptin, adipocyte complement related protein of 30 kDa (Acrp30), resistin, and perilipin. Perilipin is an intracellular neutral lipid droplet protein that is hormonally regulated. This protein is localized exclusively to the surface of lipid droplets. In response to lypotic stimuli, perilipin is phosphorylated by protein kinase A. Once activated, perilipin has inhibitory affects upon hormone-sensitive lipase (HSL), a protein that mediates the hydrolysis of triacylglycerol, the major form of stored energy in the body. Perilipin expression is limited to adipocytes and steroidogenic cells. There are currently two known isoforms, Perilipin A and B. Both of these proteins are encoded by a single-copy gene and are the result of differential splicing events.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adipophilin; adipose differentiation related protein; Adipose differentiation-related protein; ADRP; Lipid droplet-associated protein; PERI; Perilipin A; perilipin B; Perilipin-1; Perilipin-2
Gene Aliases: 6030432J05Rik; AA407157; Adfp; ADPH; Adrp; Peri; PERIA; Plin; Plin1; Plin2
UniProt ID: (Mouse) Q8CGN5, (Mouse) P43883
Entrez Gene ID: (Mouse) 103968, (Rat) 25629, (Mouse) 11520, (Rat) 298199
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support