Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Arylalkylamine N-Acetyltransferase (AANAT) is the penultimate enzyme is melatonin synthesis. Hormonal melatonin has been implicated in sleep and circadian clock function. Large changes in the abundance of AANAT are believed to be responsible for the large day/night rhythms in melatonin synthesis in the pineal gland and retina. Most research shows that AANAT activity is regulated by controlling the steady-state levels of the protein. Recent research, however, suggests that another mechanism may also play a role in controlling melatonin production without altering AANAT protein and activity. This mechanism may explain the sudden physiological increase in circulating melatonin observed at the onset of the dark period in humans.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AA-NAT; Aralkylamine N-acetyltransferase; arylakylamine N-acetyltransferase; Arylalkylamine N - acetyltransferase (Serotonin N - acetyltransferase); arylalkylamine N-acetyltransferase; Seretonin N-acetyltransferase; Serotonin acetylase; Serotonin N-acetyltransferase
Gene Aliases: AA-NAT; AANAT; DSPS; Nat-2; Nat4; SNAT
UniProt ID: (Human) Q16613, (Mouse) O88816, (Rat) Q64666
Entrez Gene ID: (Human) 15, (Mouse) 11298, (Rat) 25120
Molecular Function: acetyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support